Copper(II) Nitrate Trihydrate

Supplier: ALDON CORP SE

CC0515-100G CC0515-500G CC0515-PK/4 CC0517-CONC CC0518-500ML CC0515-100GR CC0515-500GR CC0519-500ML CC0517-500ML
470300-834EA 38 CAD
470300-834 470300-836 470300-842 470300-830 470300-832 470300-838 470300-840 470225-778 470328-168
Copper(II) Nitrate Trihydrate
Chemicals for Science Education

CAS Number: 10031-43-3
Formula Weight: 241.60
Formula: Cu(NO3)2·3H2O
Hazard Info: Toxic, Oxidizer
Density (g/mL): 2.32
Freezing Point (°C): 114.5
Solubility: Water and Alcohol
Synonyms: Copper Nitrate, Trihydrate, Copper(II) Nitrate 3-Hydrte
Shelf Life (months): 12
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR