Copper(II) Chloride Dihydrate

Supplier: ALDON CORP SE

CC0500-500G CC0500-500GR CC0500-100G CC0500-2.5KG CC0500-100GR CC0500-25GR CC0500-PK/4
470300-812EA 76.7 CAD
470300-812 470300-818 470300-826 470300-810 470300-814 470300-816 470300-820
Copper(II) Chloride Dihydrate
Chemicals for Science Education

CAS Number: 10125-13-0
Formula: CuCl2 2H2O
Density: 2.54 g/mL
Freezing Point: 100°C
Solubility: Water and Alcohol
Synonyms: Cupric Chloride Dihydrate
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR