Copper(II) Chloride Anhydrous

Supplier: ALDON CORP SE

CC0510-100G CC0510-500G CC0510-500GR CC0509-500ML CC0511-250ML CC0510-PK/4 CC0506-500ML AD-CC0507-20ML
470300-804EA 66.35 CAD
470300-804 470300-806 470300-808 470300-822 470300-824 470300-828 470225-722 470357-908
Copper(II) Chloride Anhydrous
Chemicals for Science Education

CAS Number: 7447-39-4
Formula: CuCl2
Density: 2.54 g/mL
Boiling Point: Decomposes
Freezing Point: 100 °C
Synonyms: Cupric Chloride Anhydrous


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a toxic irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR