Copper (II) Acetate Monohydrate

Supplier: ALDON CORP SE

CC0475-500G
470300-796EA 106.2 CAD
470300-796
Copper (II) Acetate Monohydrate
Chemicals for Science Education

CAS Number: 6046-93-1
Formula Weight: 199.65
Formula: Cu(O2CCH3)2·H2O
Hazard Info: Toxic
Freezing Point (°C): 115
Solubility: Water, Alcohol and Ether
Synonyms: Cupric Acetate Monohydrate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR