Copper(I) Chloride Anhydrous

Supplier: ALDON CORP SE

CC0560-100G CC0560-500G
470300-792EA 28.6 CAD
470300-792 470300-794
Copper(I) Chloride Anhydrous
Chemicals for Science Education

CAS Number: 7758-89-6
Formula Weight: 99.00
Formula: CuCl
Hazard Info: Toxic, Irritant
Density (g/mL): 3.53
Boiling Point (°C): 1366
Freezing Point (°C): 422
Solubility: Acids, Ammonia, Ether and Water
Synonyms: Cuprous Chloride Anhydrous
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR