Ward's® Prepared Contact Plates For Bacterial Recovery
Supplier: Culture Media & Supplies
CM163-RP15 CM392-RP15 CM103RP15
470175-936PK 77.8 CAD
470175-936 470175-938 470193-038
Ward's® Prepared Contact Plates For Bacterial Recovery
Educational Materials Biology Educational Materials Microbiology Learning Activities
Fast and Simple
  • No swabbing
  • Count colonies using grid pattern
  • For measuring contamination
These contact plates require no swabbing or evening. Simply remove the lid, press the raised medium against the surface being tested, replace the lid and allow to incubate. Lecithin and polysorbate 80 are added to the Total Bacterial and Yeast/Mold recovery plates to neutralize residual compounds that may be present on the material being sampled.
The number of colonies can be counted using the 10mm grid pattern on the bottom of the plates. These plates areMore Product Information