Congo Red

Supplier: ALDON CORP SE

CC0395-10G CC0405-100ML CC0409-100ML CC0410-100ML
470300-790EA 30.95 CAD
470300-790 470300-784 470300-786 470300-788
Congo Red
Chemicals for Science Education

CAS Number: 573-58-0
Formula Weight: 696.68
Formula: C32H22N6Na2O6S2
Hazard Info: Toxic
Solubility: Water and Alcohol
Synonyms: Direct Red 28, C.I. No. 22120, Congo Red Stain, Congo Red Biostain
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR