Complete Precision Air Table
Supplier: S R P INC CP
T01-001F 0011068 F15-012
470006-994EA 5804.13 CAD
470006-994 470006-996 470144-360 470200-444 470200-446 470200-448
Complete Precision Air Table
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Complete Precision Air Table helps investigate low-friction collisions.
  • Instructions with Suggested Experiments Included
  • Air Table Made of Heavy Glass
  • Air Compressor Included
  • Spark Sources Works with Six Frequencies
The recording system on this air table makes it easy to take good quantitative measurements from an air-table experiment.
A spark from the spark generator leaves a mark on the recording paper placed in the table. Students then use a protractor and centimeter scale to take measurements. The spark source can be operated at sixMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable