Ward's® Live American Cockroaches (Periplaneta americana)
Supplier: ENTOMOLOGICAL RESEARCH
876150 876151 PERIPKG50
470180-344PK 64.7 CAD
470180-344 470180-346 470180-352
Ward's® Live American Cockroaches (Periplaneta americana)
Specimens Live Specimens Animals - Living Material
Cockroaches are widely used in labs for physiological, behavioral, structural, and developmental studies.
  • Specimens come from cultures bred in the lab environment
This species is also used regularly to test the effectiveness of pesticides.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.