Cobalt(II) Nitrate Hexahydrate

Supplier: ALDON CORP SE

CC0360-100G CC0360-500G CC0360-100GR CC0357-500ML CC0358-CONC
470300-768EA 49.95 CAD
470300-768 470300-770 470300-772 470300-774 470300-776
Cobalt(II) Nitrate Hexahydrate
Chemicals for Science Education

CAS Number: 10026-22-9
Formula Weight: 291.03
Formula: Co(NO3)2·6H2O
Hazard Info: Oxidizer
Density (g/mL): 1.88
Freezing Point (°C): 55
Solubility: Most Organic Solvents
Synonyms: Cobalt Nitrate Hexahydrate, Cobaltous Nitrate Hexahydrate
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR