Cobalt(II) Chloride Hexahydrate

Supplier: ALDON CORP SE

CC0349-100G CC0349-100GR CC0349-500GR CC0349-500G CC0352-CONC CC0352-500ML CC0356-500ML CC0353-500ML
470300-750EA 45.1 CAD
470300-750 470300-754 470300-756 470300-752 470300-758 470300-760 470225-650 470225-652
Cobalt(II) Chloride Hexahydrate
Chemicals for Science Education

CAS Number: 7791-13-1
Formula: CoCl2 6H2O
Density: 1.92 g/mL
Solubility: Water, Alcohol, and Acetone
Synonyms: Cobaltous Chloride Hexahydrate, Cobalt Dichloride Hexahydrate
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR