Cobalt

Supplier: ALDON CORP SE

CC0345-100G CC0345-25G
470300-764EA 210.2 CAD
470300-764 470300-766
Cobalt
Chemicals for Science Education

CAS Number: 7440-48-4
Formula Weight: 58.93
Formula: Co
Density (g/mL): 8.9
Boiling Point (°C): 2723
Freezing Point (°C): 1493
Solubility: Nitric Acid
Synonyms: Cobalt Metal Pieces
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR