Ward's® Coal (Lignite)
Supplier: Avantor
470015-464PK 33.8 CAD
470015-464 470025-950 470025-952
Ward's® Coal (Lignite)
Specimens Rock Specimens Sedimentary Rocks
Brownish black, intermediate stage between peat and bituminous coal.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in theMore Product Information