Cladophora, General Slide
Supplier: Avantor
WARD470180-968
470180-968EA 10.45 CAD
470180-968
Cladophora, General Slide
Slides Prepared Slides
Filamentous Algae
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Branched filaments with multinucleate cells.