Citrus Cleaner
Supplier: Aldon
CC0343-250
470006-770EA 38.7 CAD
470006-770
Citrus Cleaner
Cleaning Agents Detergents and Surface Cleansers
Squeeze bottle cleans grease, marker, adhesive, and wax from any surface.
Oil based cleaner not intended for use with lenses. Recommended by microscope repair specialist Joyce Dorsey for cleaning microscope bodies. 250 mL bottle.