Citric Acid Monohydrate

Supplier: ALDON CORP SE

CC0335-500G CC0335-100G CC0335-500GR CC0331-50ML
470300-742EA 43 CAD
470300-742 470300-740 470300-744 470225-822
Citric Acid Monohydrate
Chemicals for Science Education

CAS Number: 5949-29-1
Formula: C6H8O7·H2O
Density: 1.542 g/mL
It Loses H2O at 100°C
Solubility: Water, Alcohol and Ether
Synonyms: Propane Tricarboxylic Acid
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR