Citric Acid Anhydrous

Supplier: ALDON CORP SE

CC0337-100G CC0337-500G
470300-734EA 21.9 CAD
470300-734 470300-736
Citric Acid Anhydrous
Chemicals for Science Education

CAS Number: 77-92-9
Density: 1.665 g/mL
Freezing Point: 153 °C
Solubility: Water, Alcohol and Ether
Synonyms: Propane Tricarboxylic Acid
Shelf Life: 12 Mouths


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR