Ward's® Live Drosophila melanogaster - Chromosome III Mutants
Supplier: Ward's Science
CALSEPIA CALSEPEB 876583
470176-758EA 17.3 CAD
470176-758 470176-762 470176-772 470180-428 470180-430 470180-446
Ward's® Live Drosophila melanogaster - Chromosome III Mutants
Specimens Live Specimens Animals - Living Material
Drosophila melanogaster, the common fruit fly, is widely used in genetic studies. They express a number of mutations such as eye, wing, and body mutations.
Students can study sex-linked, recessive, and dominant mutations. Drosophila also make an excellent live food for mantises, small reptiles and amphibians. Drosophila demonstrates complete metamorphosis with egg, larva, pupa, and adult stages in its life cycle.
The pupae will usually appear within ten days of the ship date. During very cold weather, adult flies may die but eggs and larvae usually survive. Allow cultures to come toMore Product Information