Chlorophenol Red
Supplier: ALDON CORP SE
CC0315-100ML
CC0314-1G
470300-706EA
55
CAD
470300-706
470300-708
Chlorophenol Red
Chemicals for Science Education
CAS Number: 4430-20-0
Formula: C19H12Cl2O5S
Formula Weight: 423.27
Shelf Life (months): 36
Solubility: Water
Storage: Green
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...