Chloroform

Supplier: ALDON CORP SE

CC0275-500ML CC0275-1LR CC0275-500MLR
470045-588EA 118.2 CAD
470045-588 470300-702 470300-704
Chloroform
Chemicals for Science Education

CAS Number: 67-66-3
Formula Weight: 119.38
Formula: CHCl3
Hazard Info: Toxic, Narcotic
Density (g/mL): 1.48
Boiling Point (°C): 61.1
Solubility: Water
Synonyms: Trichloromethane
Shelf Life (months): 36
Storage: Blue


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR