Charcoal

Supplier: ALDON CORP SE

CC0250-50G CC0260-500G CC0250-500G CC0265-500G CC0260-500GW AD-CC0256-1LB CC0251-1KG
470300-658EA 18.1 CAD
470300-658 470300-688 470300-690 470300-692 470300-694 470220-578 470303-578
Charcoal
Chemicals for Science Education

CAS Number: 7440-44-0
Formula Weight: 12.01
Formula: C
Hazard Info: Flammable
Density (g/mL): 0.4-0.7
Shelf Life (months): 36
Storage: Green/Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR