Hanging Cell Models
Supplier: SCIENCE INTERACTIVE GROUP
B 140 B 142
470002-684EA 698.45 CAD
470002-684 470000-002
Hanging Cell Models
Models Molecular Biology Models
For Demonstration And Display
These high visibility large models are vacuum formed sheets of heavy plastic, molded to show in relief all the known organelles and inclusions of these two important cells. Twenty structures are included in the animal cell; twenty-three in the plant cell. All are identified by the accompanying key cards. Hand painted in brilliant color and are fitted with an easel back which permits the model to sit upright on a surface of hang on a wall with equal ease.