Preserved Cats
Supplier: Avantor
53192 140.1038.1 53193 140.1040.1 140.1008.1 140.1022.1 140.1014.1 DEFAULT VPN
470154-100EA 216.4 CAD
470154-100 470154-104 470154-106 470154-110 470154-112 470154-116 470154-120 470154-126 CA470315-546 CA470315-758
Preserved Cats
Specimens Preserved Specimens Preserved Animal Materials
Standard formalin preserved cats, packed in vacuum-sealed bags. One cat per bag.
  • Vertebrate anatomy
  • Mammalian anatomy
  • High quality preservation
Cats, Felis species are expertly preserved and presented in an extended position for anatomical correctness and ease of study. Our latex injected cats are injected with elastic latex with bright, contrasting colors, to ensure ease of dissection. Plastic storage bags and waterproof ID tags are provided free with each cat.
Caution: Cats are sourced from pounds and shelters. All shelters are required to retainMore Product Information