Carmine

Supplier: ALDON CORP SE

CC0190-1G CC0190-10G
470300-666EA 24.5 CAD
470300-666 470300-668
Carmine
Chemicals for Science Education

CAS Number: 1390-65-4
Formula Weight: 492.39
Formula: C22H20O13/Al/Ca lake
Density (g/mL): >1.0
Boiling Point (°C): Decomposes
Freezing Point (°C): 120
Synonyms: C.I. Natural Red 5, C.I. No 75470, Alum Lake of Carminic Acid
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR