Carbon Rod Electrode

Supplier: Shiv Dial Sud & Sons

P50070/5 P50070/55
470149-046CS 1752.1 CAD
470149-046 470149-520
Carbon Rod Electrode
Chemicals for Science Education

CAS Number: 7440-44-0
Formula Weight: 12.01
Formula: C
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR