Carbol Fuchsin Stain
Supplier: ALDON CORP SE
CC0165-100ML
CC0163-10G
470300-652EA
20.55
CAD
470300-652
470300-654
Carbol Fuchsin Stain
Chemicals for Science Education
CAS Number: 4197-24-4
Formula: C21H22CIN3
Shelf Life: 36 Months
Storage: Blue/Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...