Capillarity Apparatus
Supplier: VWR International
PH0202C
470014-976EA 34.6 CAD
470014-976
Capillarity Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Capillarity Apparatus is intriguing demonstration of molecular attraction.
  • Four Glass Tubes
  • Metal Bass
  • Length: 5½"
  • Width: 4¾"
Adding colored water to the reservoir tubes of the apparatus shows how water ascends into spaces by capillary action. The water levels will vary in the four differently-sized glass tubes, rising above the reservoir level in the narrower tubes.
The glass unit is firmly supported in a metal base and measures 5½ x 4¾".