Candida albicans
Supplier: VWR
WARD470179-636
470179-636EA 22.05 CAD
470179-636
Candida albicans
Specimens Live Specimens Fungi - Living Material
  • Biosafety Level 2
  • Canada: Containment Level 2 - PHAC License required
Deuteromycete. Causes thrush, candidiasis. Multiplies by budding. Forms germ-tubes in serum cultures. Incubation temperature 37°C. Potato dextrose agar growth medium. Note: Pathogen.