Calcium Sulfate Hemihydrate

Supplier: ALDON CORP SE

CC0155-500G CC0155-1KG CC0155-2.5KG
470300-646EA 17.6 CAD
470300-646 470300-642 470300-644
Calcium Sulfate Hemihydrate
Chemicals for Science Education

CAS Number: 26499-65-0
Formula Weight: 145.15
Formula: CaSO4·12H2O
Density (g/mL): 2.8
Solubility: Water
Synonyms: Plaster of Paris, Dried Gypsum
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR