Calcium Sulfate Dihydrate

Supplier: ALDON CORP SE

CC0150-500G CC0150-500GR
470300-638EA 56.65 CAD
470300-638 470300-640
Calcium Sulfate Dihydrate
Chemicals for Science Education

CAS Number: 10101-41-4
Formula Weight: 172.10
Formula: CaSO4·2H2O
Density (g/mL): 2.32
Freezing Point (°C): 128
Solubility: Water
Synonyms: Calcium Sulfate 2-Hydrate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR