Calcium Hydroxide

Supplier: ALDON CORP SE

CC0095-100G CC0095-500G CC0090-500GR CC0101-500ML CC0100-500ML
470045-514EA 20.95 CAD
470045-514 470300-588 470300-590 470225-678 470225-774
Calcium Hydroxide
Chemicals for Science Education

CAS Number: 1305-62-0
Formula Weight: 74.10
Formula: Ca(OH)2
Hazard Info: Irritant, Corrosive
Density (g/mL): 2.24
Boiling Point (°C): as CaO 579
Freezing Point (°C): as CaO 2580
Solubility: Water, Glycerol, and Acids
Synonyms: Hydrated Lime, Slacked Lime, Caustic Lime, Calcium Hydrate
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR