Calcium Chloride

Supplier: ALDON CORP SE

CC0075-500G CC0075-2.5KG CC0080-500GR
470300-556EA 21.35 CAD
470300-556 470300-558 470300-568
Calcium Chloride
Chemicals for Science Education

CAS Number: Mixture
Formula: CaCl2
Freezing Point: 772 °C
Shelf Life: 12 Months
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR