Calcium Carbide
Supplier: ALDON CORP SE
CAS Number: 75-20-7
Formula: CaC2
Density: 2.20 g/mL
Freezing Point: 1700 to 2300°C
Solubility: Water
Synonyms: Acetylene Oxygen, Carbide
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: The product is flammable and corrosive.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...