Calcium Acetate Monohydrate

Supplier: ALDON CORP SE

CC0040-500ML CC0038-100G CC0038-500G
470300-526EA 29.55 CAD
470300-526 470300-528 470300-530
Calcium Acetate Monohydrate
Chemicals for Science Education

CAS Number: 5743-26-0
Formula Weight: 176.18
Formula: Ca(CH3CO2)2·H2O
Density (g/mL): 0.3
Boiling Point (°C): Decomposes
Synonyms: Lime Acetate, Calcium Diacetate
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR