Calcium

Supplier: ALDON CORP SE

CC0035-100G CC0035-25G CC0035-500G CC0035-PK/4
470300-598EA 91.7 CAD
470300-598 470300-600 470300-596 470300-602 470300-604 470300-606 470300-608
Calcium
Chemicals for Science Education

CAS Number: 7440-70-2
Formula: Ca
Density: 1.54 g/mL
Boiling and Freezing Point: 1440°C, 842°C
Solubility: Acid
Synonyms: Calcium Metal
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a flammable irritant.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR