Caffeine

Supplier: ALDON CORP SE
CC0025-25G CC0025-100G CC0025-10G
470300-524EA 20.45 CAD
470300-524 470300-522 470300-520
Caffeine
Chemicals for Science Education
CAS Number: 58-08-2
Formula Weight: 194.20
Formula: C8H10N4O2
Hazard Info: Toxic
Density (g/mL): 1.23
Boiling Point (°C): 178
Freezing Point (°C): 238
Solubility: Chloroform, Water and Alcohol
Synonyms: 1,3,7-Trimethylxanthine
Shelf Life (months): 12
Storage: Blue

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR