Butylated Hydroxy Toluene

Supplier: ALDON CORP SE

IX0240-100G IX0240-500G
470300-506EA 41.4 CAD
470300-506 470300-508
Butylated Hydroxy Toluene
Chemicals for Science Education

CAS Number: 128-37-0
Formula Weight: 220.35
Formula: C15H24O
Hazard Info: Flammable, Toxic, Irritant
Density (g/mL): 0.899
Boiling Point (°C): 265
Freezing Point (°C): 69.44
Synonyms: 2,6-Di-Tert-Butyl-4-Methylphenol, BHT
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR