Ward's® Build Your Own Collection Kits
Supplier: Avantor
470022-432KT 127.65 CAD
470022-432 470022-434 470022-436 470022-438 470022-440
Ward's® Build Your Own Collection Kits
Specimens Rock Specimens Rock Collections
Everything You Need to Organize Your Specimens
You supply the minerals, rocks, and fossils; we supply the rest. Each kit contains ten compartmented white plastic trays, ten sturdy collection boxes, ten sets of printed numbers for labeling samples, glue, and a label applicator.
Collection Kits with 12-Compartment Trays
Box size: 7 1/4"L x 7 1/4"W x 1 1/2"H; tray cavity size: 2"L x 1 1/2"W.
Collection Kit with 16-Compartment Trays
Box size: 7 1/4"L x 7 1/4"W x 1 1/2"H; tray cavity size: 1 1/2"L x 1 1/2"W.
Collection Kit with 24-Compartment Trays
Box size: 13 1/4"L x 10 1/4"W x 1 3/4"H; tray cavity size: 2 1/4"L x 1 1/8"W.
Collection Kit with 36-Compartment Trays
Box size: 13 1/4"L x 10 1/4"W x 1 3/4"H; tray cavity size: 2"L x 1 1/2"W.
Collection Kit with 45-Compartment Trays
Box size: 16 1/2"L x 13"W x 1 3/4"H; tray cavity size: 2 1/2"L x 1 3/4" W.
Collection Kit with 50-Compartment Trays
Box size: 16 1/2"L x 13"W x 1 3/4"H; tray cavity size: 2 1/4"L x 1 3/8"W.