Buffer Solution
Supplier: ALDON CORP SE
BB0410-1L
BB0403-1L
BBR404-500ML
BB0404-1L
BBY407-500ML
BB0407-1L
BB0411-500ML
BBB410-500ML_B
BB0402-1L
BB0403-500ML
BB0405-1L
BB0405-500ML
BB0406-1L
BB0406-500ML
BB0408-1L
BB0408-500ML
BB0409-1L
BB0409-500ML
BB0412-500ML
BB0410-500ML
470300-460EA
37.2
CAD
470300-460
470300-468
470300-474
470300-476
470300-488
470300-490
470300-502
470300-464
470300-466
470300-472
470300-480
470300-482
470300-484
470300-486
470300-494
470300-496
470300-498
470300-500
470300-504
470300-462
Buffer Solution
Buffers
Cell Culture Buffers
Buffer solution has a shelf life of 24 months
- Density: Approximately 1 g/mL
- Boiling Point: 100°C
- Freezing Point: 0
- Shelf Life: 24 Months
- Storage: Green
Its density is 1 g/mL.
Buffer solution has a boiling point of 100°C and a freezing point of 0°C.
Caution:
This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...