Buffer Capsules

Supplier: MICRO ESSENTIAL LABORATORY I
270-PH10 270-PH6.0 270-PH7.0 270-PH3.0 270-PH09 270-PH12-B 270-PH8.0 270-PH5.0 270-PH4.0 270-PH11 270-PH2.0
470158-204CS 90.4 CAD
470158-204 470158-194 470158-172 470158-182 470158-160 470158-208 470158-176 470158-164 470158-186 470158-168 470158-200
Buffer Capsules
Buffers Cell Culture Buffers
Shelf Life (months): 36
Storage: Green

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR