Bromothymol Blue

Supplier: ALDON CORP SE

BB0220-30ML BB0220-500ML BB0222-500ML BB0220-100ML BB0225-500ML BB0221-30ML BB0215-1GR BB0215-5GR
470300-440EA 18.65 CAD
470300-440 470300-448 470300-450 470300-442 470300-446 470300-452 470300-454 470300-456 470300-458
Bromothymol Blue
Chemicals for Science Education

CAS Number: 34722-90-2
Formula: C27H27Br2O5SNa
Boiling Point: Decomposes
Solubility: Water
Synonyms: Bromothymol Blue pH Indicator
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR