Bromocresol Purple

Supplier: ALDON CORP SE

BB0199-500ML BB0199-100ML BB0195-1G
470300-426EA 17.85 CAD
470300-426 470300-428 470300-430
Bromocresol Purple
Chemicals for Science Education

CAS Number: 62625-30-3
Formula Weight: 562.20
Formula: C21H15Br2O5S·Na
Boiling Point (°C): Decomposes
Solubility: Water
Synonyms: Bromocresol Purple, pH Indicator
Shelf Life (months): 36
Storage: Green/Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR