Graduated Cylinders with Plastic Base
Supplier: United Scientific Supplies
CY137-100 CY137-10 CY137-25 CY137-50 CY137-50 CY137-250 CY137-1000 CY137-500
470121-934PK 219 CAD
470121-934 470188-950 470188-952 470188-954 470188-955 76531-552 470349-758 470349-806
Graduated Cylinders with Plastic Base
Cylinders Measuring Cylinders
Durable Borosilicate Glass Construction.
  • Graduations permanently fused into glass
  • Hexagonal base and bumper guard add protection for all sizes
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing.
Order Now
SPECIFICATIONS
Form Tall
Environmentally Preferable