Boreal2 Research Microscopes, AP Series
Supplier: VWR
Boreal2
SK233
470014-534EA 2543.55 CAD
470014-534
Boreal2 Research Microscopes, AP Series
Microscopes
Research microscopes with centerable Abbe Condenser, ACS objectives, and Koehler illumination for advanced microscopy.
  • 4×, 10×, 40×, 100× ASC objectives
  • Built-In mechanical stage
  • Full size: 14⅝" H
  • Inclined trinocular head
  • Boreal lifetime warranty
Revolutionize how you teach microscopy to advanced students with this research-grade microscope offering superior images. Using accessories available separately, this innovative microscope offers phase contrast, darkfield, or other specialized imaging techniques. ACS objectives provide images that rival semi-plan objectives and the large, heavy body ensures a stable image. Both 40× and 100× objectives are sealed for protection against stain and oil, while other tamper-proof features include a locked mechanical stage and recessed set screws for the eyepieces and stage stops. A Seidentopf trinocular head offers comfortable viewing over long sessions and it adjusts without affecting focus. Precision movements of the stage and fine focus also help to pinpoint detailed structures. This compound model offers white, bright cool illumination, a removable cord rack for easy storage, and a lifetime warranty.