Borax

Supplier: ALDON CORP SE

SS0338-100ML SS0335-500ML
470300-404EA 11.4 CAD
470300-404 470300-406
Borax
Chemicals for Science Education

CAS Number: 1303-96-4
Formula Weight: Mixture
Density: Approximately 1 g/mL
Boiling and Freezing Point: Approximately 100°C, Approximately 0°C
Solubility: Water
Synonyms: Sodium Borate Water Solution
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR