Bone Clones® Hominids Skull Series
Supplier: BONE CLONES HOLDINGS
BH-025 BH-023 BH-020 BH-021A BH-008 BH-016 BH-013 BH-004 BH-012 BH-018
470029-216EA 368.55 CAD
470029-216 470029-218 470029-220 470029-222 470029-224 470029-226 470029-228 470029-230 470029-232 470029-234 470029-238
Bone Clones® Hominids Skull Series
Models Paleontology Models
The most complete series of human ancestry
  • Buy individually or bundle and save
  • Accurate replicas are great for human biology, evolutionary biology, and athropology courses
Each hominid was carefully researched and re-created in resin from casts that were made from the original fossils, the latest literature, and photographs, then color-coded to distinguish the original fossil remains from the reconstructed parts.
Aegyptopithecus is believed to be an ancestor of Old World monkeys and hominids. Proconsul africanus lived in Africa approximately 14 -23More Product Information