Prepared Blood Agar Media
Supplier: CULTURE MEDIA &SUPPLIES INC SE
880070
880901
470180-642CS
560.9
CAD
470180-642
470180-680
Prepared Blood Agar Media
Media for Microbiology
Dehydrated Media for Microbiology
Tryptic Soy Agar base with 5% Sheep Blood
- Excellent for cultivation of fastidious organisms
- Plate your organism onto Blood agar plates to test for hemolytic reaction