Blocks and Refraction Plates
Supplier: Abrisa-USPG Industrial Glass
1-58 PH0566D PH0568-13 PH0568E PH0568E
470111-286EA 17.89 CAD
470111-286 470110-632 470110-644 470110-646 470110-648
Blocks and Refraction Plates
Educational Materials Physics Educational Materials Optics Learning Activities
Parallel-Sided Blocks for Ray Optics
Ideal for refraction and internal reflection studies as well as index of refraction measurements, these clear blocks are free of bubbles and streaks and have polished surfaces. Acrylic blocks and refraction plates have frosted ends, glass block is completely clear.
Order Now
SPECIFICATIONS
Environmentally Preferable