Biuret Reagent Solution

Supplier: ALDON CORP SE

BB0150-1L BB0150-100ML BB0150-500ML BB0145-50G
470300-394EA 30.75 CAD
470300-394 470300-396 470300-398 470300-392
Biuret Reagent Solution
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Density: 1.0 g/mL
Boiling and Freezing Point: Approximately 100°C, Approximately 0°C
Synonyms: Biuret Solution, Biuret Reagent
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This item is corrosive.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR