Bismuth Nitrate Pentahydrate

Supplier: ALDON CORP SE

BB0143-100GR BB0143-25GR
470300-386EA 142.05 CAD
470300-386 470300-388
Bismuth Nitrate Pentahydrate
Chemicals for Science Education

CAS Number: 10035-06-0
Formula Weight: 485.1
Formula: Bi(NO3)3·5H2O
Hazard Info: Oxidizer, Flammable, Irritant
Density (g/mL): 2.830
Boiling Point (°C): 80
Freezing Point (°C): 30
Solubility: Nitric Acid, Alcohol and Acetone
Synonyms: Bismuth(III) Nitrate 5-Hydrate, Bismuth Trinitrate, Bismuth Nitrate
Shelf Life (months): 12
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR